Learn More
Medchemexpress LLC Brinzolamide | 138890-62-7 | 99.9% | 50 MG

Supplier: Medchemexpress LLC HYB058850MG
Brinzolamide is a selective carbonic anhydrase II inhibitor with an IC50 value of 3.2 nM. It reduces intraocular pressure (IOP) by inhibiting ciliary CA-II and decreasing atrial fluid secretion, and can be used in glaucoma disease research.
- Selective carbonic anhydrase II inhibitor
- IC50 value of 3.2 nM
- Reduces intraocular pressure (IOP) by inhibiting ciliary CA-II
- Can be used in glaucoma disease research
- Molecular weight: 383.51
- Formula: C12H21N3O5S3
- Appearance: Solid
- Color: Light yellow to yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.