Learn More
Medchemexpress LLC Brinzolamide | 138890-62-7 | 99.9% | 5 MG
Supplier: Medchemexpress LLC HYB05885MG
Brinzolamide (AL-4862) is a selective carbonic anhydrase II inhibitor with an IC50 value of 3.2 nM. This compound reduces intraocular pressure (IOP) by inhibiting ciliary CA-II and decreasing atrial fluid secretion, making it suitable for glaucoma disease research.
- Selective carbonic anhydrase II inhibitor
- Reduces intraocular pressure (IOP)
- Inhibits ciliary CA-II and decreases atrial fluid secretion
- Suitable for glaucoma disease research