Learn More
Cole-Parmer BRISKHEAT DPCS10 FLEXIBLE 5-GA
Supplier:  Cole-Parmer 0310673
Encompass_Preferred
BriskHeat DPCS10 Flexible 5-Gal Poly Drum Heater with Thermostat; 50-160F, 120V
Catalog No. NC2132178