Learn More
Medchemexpress LLC Britannin | 33627-28-0 | 99.9% | 366.41 | C19H26O7 | 10 MG

Supplier: Medchemexpress LLC HYN300510MG
Britannin is a small-molecule research reagent reported to inhibit NLRP3 inflammasome activation and to induce autophagy; intended for laboratory research use only.
- Potent NLRP3 inhibitor with reported IC50 3.63 μM.
- High purity (99.93%).
- Molecular formula C19H26O7; molecular weight 366.41 g/mol.
- White to off-white solid suitable for biochemical and cellular assays.
- Storage: store at 4°C and protect from light; solutions stable at -80°C (6 months) or -20°C (1 month).
- Available in small milligram quantities and DMSO solution formats for ready use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.