Learn More
TARGETMOL CHEMICALS INC BRIVANIB ALANINATE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T25765MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Brivanib Alaninate (BMS-582664) is the alaninate salt of a vascular endothelial growth factor receptor 2 (VEGFR2) inhibitor with potential antineoplastic activity. Brivanib strongly binds to and inhibits VEGFR2 a tyrosine kinase receptor expressed almost exclusively on vascular endothelial cells; inhibition of VEGFR2 may result in inhibition of tumor angiogenesis inhibition of tumor cell growth and tumor regression. purity: 99%
Catalog No. 50-257-7090