Learn More
Selleck Chemical LLC Brivanib Alaninate (BMS-582664) 10mg 649735-63-7
SDP
Supplier:  Selleck Chemical LLC S113810MG
Encompass_Preferred
Brivanib alaninate (BMS-582664) is the prodrug of BMS-540215, an ATP-competitive inhibitor against VEGFR2 with IC50 of 25 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-623-5