Learn More
Abcam BRL-50481, Selective PDE7 inhibitor, 50MG
Supplier:  Abcam AB14170550MG
Encompass_Preferred
MW 244.27 Da, Purity >98%. Selective, competitive PDE7 inhibitor (IC₅₀ values are 0.15 and 12.1 μm for PDE7A and PDE7B respectively). Active in vivo and in vitro. Potentiates the anti-inflammatory effect of PDE4 inhibitor rolipram (ab120031). Cell and blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6474