Learn More
Abcam BRL 52537 hydrochloride, kappa-opioid agonist, 50MG
Supplier:  Abcam AB12066950MG
Encompass_Preferred
MW 391.77 g/mol, Purity >99%. Highly selective and potent κ-opioid agonist (Ki = 0.24 nM). 25 times more potent than morphine. Neuroprotective effects seen in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7365