Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask-Grad,4000mL Narrow mouth
SDP
Supplier:  Bellco Glass, Inc. 250404000
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Size: 4l Stopper size: 10. OD X OAH (MM):205 x 355. CASE QTY:3
Catalog No. 50-194-5445