Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask-Grad,500mL Narrow mouth
SDP
Supplier:  Bellco Glass, Inc. 250400500
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Size: 500ml Stopper size: 7. OD X OAH (MM):100 x 183. CASE QTY:12
Catalog No. 50-194-5441