Learn More
Bellco Glass, Inc. Break Resistant Lip Media/Disp Flask WHC,1000mL Graduated
SDP
Supplier:  Bellco Glass, Inc. 253001000
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Media storage and dispensing flask with bottom hose connection, accepts 1/2" tubing. Graduated to approximate volumes and the heavy duty break resistant neck provides an excellent fit for rubber stoppers. Not for vacuum use.. MADE IN THE USA Size: 1L Dimensions: 128x223mm
Catalog No. 50-194-5505