Learn More
Bellco Glass, Inc. Break Resistant Lip Shake Flask W/3 Baffs,1L
SDP
Supplier:  Bellco Glass, Inc. 254101000
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with rigidly controlled baffle angles and depths. The 3 side baffles are used to promote optimal oxygen transfer while reducing vortex formation. Has a break resistant lip to resist chipping and is designed for use with orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 1LDimensions: 128x220mmCase Quantity:2541-01000: 6
Catalog No. 50-194-5520