Learn More
Chemglass Life Sciences Flask, Shake, 2000mL, Baffled, Plain Top, Approx OD x Height (mm): 155 x 270
SDP
Supplier:  Chemglass Life Sciences CLS204007
Encompass_Preferred
Flask, Shake, 2000mL, Baffled, Plain Top, Approx OD x Height (mm): 155 x 270. Baffle indents on the lower outside edge of the flask to help oxygen transfer to the culture medium when used on an orbital shaker.
Catalog No. 50-121-4620