Learn More
Bellco Glass, Inc. Break Resistant Lip X-Dp BaffleFlask,1000mL
SDP
Supplier:  Bellco Glass, Inc. 254201000
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Break resistant lip to prevent chipping.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 1LDimensions: 128x220mmCase Quantity: 6Baffle Dimensions: 10mm x60°
Catalog No. 50-194-5525