Learn More
Bellco Glass, Inc. Break Resistant Lip X-Dp BaffleFlask,2000mL
SDP
Supplier:  Bellco Glass, Inc. 254202000
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Break resistant lip to prevent chipping.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 2LDimensions: 158x290mmCase Quantity: 12Baffle Dimensions: 6mm x35°
Catalog No. 50-194-5526