Learn More
Chemglass Life Sciences Flask, Shake, 250mL, Deep Baffles, Plain Top, Approx OD x Height (mm): 80 x 135
SDP
Supplier:  Chemglass Life Sciences CLS204202
Encompass_Preferred
Flask, Shake, 250mL, Deep Baffles, Plain Top, Approx OD x Height (mm): 80 x 135. Extra deep baffle indents on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker.
Catalog No. 50-121-4721