Learn More
Medchemexpress LLC BRM/BRG1 ATP inhibitor-1 | 2270879-17-7 | 99.7% | 100 MG

Supplier: Medchemexpress LLC HY119374100MG
BRM/BRG1 ATP Inhibitor-1 is a potent, allosteric small-molecule inhibitor of the ATPase activities of BRM (SMARCA2) and BRG1 (SMARCA4), supplied as a research reagent for biochemical and cellular studies of SWI/SNF chromatin-remodeling complexes.
- Targets BRM and BRG1 ATPase activities.
- Demonstrates IC50 <0.005 μM for BRM and BRG1 ATPases.
- Molecular weight 318.27 g/mol.
- Chemical formula C11H9F3N4O2S.
- Purity approximately 99.7%.
- Powder storage at -20°C; in solvent store at -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.