Learn More
Selleck Chemical LLC BRM/BRG1 ATP Inhibitor-1
SDP
Supplier:  Selleck Chemical LLC E0111100MG
Encompass_Preferred
BRM/BRG1 ATP Inhibitor-1 (Compound 14) is an orally active inhibitors of Brahma Homolog (BRM)/SMARCA2 (BRG1) with IC50s below 0 005 M
Catalog No. 50-313-6650