Learn More
Medchemexpress LLC Brm/brg1 atp inhibitor-2 | 2368900-77-8 | 99.7% | C20H20N4O2S2 | 50 MG

Supplier: Medchemexpress LLC HY14594650MG
BRM/BRG1 ATP Inhibitor-2 is an ATPase inhibitor for the research of BAF-related disorders. It has a molecular weight of 412.53 and appears as a white to off-white solid. It is soluble in DMSO at 100 mg/mL. Stock solutions can be stored at -80°C for 6 months or -20°C for 1 month.
- For research use only
- White to off-white solid appearance
- Soluble in DMSO
- Powder stable for 3 years at -20°C
- Solutions stable for 6 months at -80°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.