Learn More
Medchemexpress LLC BRM/BRG1 ATP inhibitor-2 | 2368900-77-8 | MFCD34676590 | 99.7% | 412.53 g·mol⁻¹ | C20H20N4O2S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1459465MG
Encompass_Preferred
BRM/BRG1 ATP Inhibitor-2 is a small-molecule inhibitor of the ATPase activity of BRG1 and BRM, core ATPase subunits of the SWI/SNF (BAF) chromatin remodeling complex. It is provided for research use to support biochemical and cell-based investigations into BAF-related disorders and chromatin remodeling mechanisms.
  • Potent inhibition of BRG1 and BRM ATPase activity.
  • High chemical purity suitable for biochemical assays.
  • Soluble in common organic solvents for stock preparation.
  • Suitable for in vitro and preclinical cell-based studies.
  • Provides a tool to study chromatin remodeling and disease mechanisms.
Catalog No. 50-002-20974