Learn More
Alkali Scientific Alkali Scientific™ Broad Range dsDNA Quantification Kit, 100 assays
SDP
Supplier:  Alkali Scientific SQ1000BR100
Encompass
Broad Range dsDNA Assay KitThe Alkali Scientific Broad Range kit provides an accurate method for quantitation of dsDNA samples for a broad range quantity of DNA. Depending on sample volume, the assay kit is designed to be accurate for initial DNA sample concentrations of 0.2 to 2,000 ng/µL, providing a detection range of 4−2,000 ng
Catalog No. 50-302-9580