Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eMolecules​ Broadpharm Amino-PEG9-alcohol, 98% Purity, 413.5 MW, CAS 15332-95-3

Supplier:  eMolecules​ 563055158

Encompass_Preferred

Amino-PEG9-alcohol is a PEG linker containing amine and hydroxyl moieties. The hydrophilic PEG spacer increases the water solubility of a compound in aqueous media. The amine group is reactive with carboxylic acids, activated NHS esters, and carbonyls (ketone, aldehyde). The hydroxyl group can react to allow for further derivatization of the compound.

  • Contains amine and hydroxyl moieties
  • Hydrophilic PEG spacer increases water solubility
  • Amine group is reactive with carboxylic acids, activated NHS esters, and carbonyls
  • Hydroxyl group allows for further derivatization
  • Available for GMP-grade inquiries
  • Soluble in water, DMSO, DCM, and DMF
  • Store at -20°C
  • 98% purity

Catalog No. NC3973188


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.