Learn More
ATCC Brochothrix thermosphacta
Supplier:  ATCC 11509
Encompass
Bacterial type strain isolated from fresh pork sausage. This strain is known to produce the restriction endonuclease MthI and has applications as a bacteriophage host.
Catalog No. 50-238-2267