Learn More
Medchemexpress LLC HY-108689 5mg Medchemexpress, Broflanilide CAS:1207727-04-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086895MG
Encompass_Preferred
Medchemexpress, HY-108689 5mg Broflanilide CAS:1207727-04-5 Broflanilide is a potential insecticide and metabolized to Desmethyl-Broflanilide, which is a potent antagonist at the insect resistant-to-dieldrin (RDL) GABA Receptor, and inhibits S. litura RDL GABAR, with an IC50 value of 1.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0750