Learn More
Cayman Chemical BrombuterolhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3359210 MG
Encompass_Preferred
A β2-AR agonist; has been used as an illicit feed additive to increase the muscle-to-fat ratio of livestock
Catalog No. 50-294-6674