Learn More
Hach Company Bromcresol Green-Methyl Red Indicator Solution, 100 mL MDB
Supplier:  Hach Company 2329232
Encompass
pH indicator for alkalinity determination by titration. APHA formulation 100 mL Marked Dropping Bottle.
Catalog No. 50-199-5814