Learn More
Cayman Chemical Bromfenac-d4sodIum salt 1mg
SDP
Supplier:  Cayman Chemical 265981 MG
Encompass_Preferred
An internal standard for the quantification of bromfenac by GC- or LC-MS
Catalog No. 50-293-4800