Learn More
Selleck Chemical LLC Bromfenac Sodium S4248-1g
SDP
Supplier:  Selleck Chemical LLC S42481G
Encompass_Preferred
Bromfenac Sodium (AHR 10282R Bromsite Bromday Prolensa Xibrom) is a nonsteroidal anti-inflammatory drug (NSAID) which has anti-inflammatory activity and may block prostaglandin synthesis by inhibiting cyclooxygenase 1 and 2
Catalog No. 50-310-8641