Learn More
Apexbio Technology LLC Bromfenac Sodium 91714-93-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B16845.1
Encompass_Preferred
Bromfenac Sodium (CAS 91714-93-1) is a nonsteroidal anti-inflammatory small molecule compound characterized primarily by its capacity to inhibit cyclooxygenase enzymes (COX-1 and COX-2) By targeting these enzymes Bromfenac Sodium disrupts prostaglandin biosynthesis subsequently modulating inflammatory pathways and responses In biomedical research Bromfenac Sodium serves as a useful pharmacological tool to investigate inflammation signaling pathways and prostaglandin-mediated processes in various experimental models
Catalog No. 50-000-61970