Learn More
Medchemexpress LLC Bromfenac sodium | 91714-93-1 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1888A100MG
Encompass_Preferred
Bromfenac sodium is a potent and orally active inhibitor of COX enzymes, specifically targeting COX-1 and COX-2. It is primarily utilized in ocular inflammation research due to its significant anti-inflammatory properties.
  • Potent and orally active COX inhibitor with low IC50s
  • Inhibits transforming growth factor-β2-induced epithelial-mesenchymal transition
  • Suppresses transforming growth factor-β2-induced cell migration
  • Demonstrates significant anti-inflammatory activity in various animal models
  • More potent than indomethacin in blocking erythema
  • Reduces paw volume in animal studies
  • Blocks abdominal constriction to ACh challenge
  • Partially reduces corneal staining in specific keratoconjunctivitis sicca models
Catalog No. 50-004-09668