Learn More
Apexbio Technology LLC Bromfenac sodium hydrate 120638-55-3 10mg
SDP
Supplier:  Apexbio Technology LLC B878610
Encompass_Preferred
Bromfenac sodium hydrate (CAS 120638-55-3) is an orally active inhibitor of cyclooxygenase enzymes with potent inhibitory activity against both COX-1 and COX-2 isoforms In vitro assays have demonstrated IC50 values of approximately 5 56 nM against COX-1 and 7 45 nM against COX-2 Its mechanism involves blocking prostaglandin synthesis consequently reducing inflammatory mediator levels Bromfenac sodium hydrate is commonly employed in biomedical research addressing ocular inflammation pain and related inflammatory processes
Catalog No. 50-000-64327