Learn More
Medchemexpress LLC Bromfenac sodium hydrate | 120638-55-3 | 5MG
SDP
Supplier:  Medchemexpress LLC HYB1888B5MG
Encompass_Preferred
Bromfenac sodium hydrate, also known as Bromfenac monosodium salt sesquihydrate, is a potent and orally active inhibitor of cyclooxygenase (COX) enzymes, specifically COX-1 and COX-2 (IC50 values of 5.56 nM and 7.45 nM, respectively). Primarily utilized in ocular inflammation research, this compound exhibits high purity and appears as a light yellow to yellow solid. It demonstrates excellent solubility in both DMSO and water.
  • Potent and orally active COX inhibitor.
  • Utilized in ocular inflammation research.
  • High purity of 99.94%.
  • Appears as a light yellow to yellow solid.
  • Soluble in DMSO and water (≥ 100 mg/mL).
  • Demonstrates anti-inflammatory activity and inhibition of epithelial-mesenchymal transition.
Catalog No. 50-004-11396