Learn More
TARGETMOL CHEMICALS INC Bromfenac sodium hydrate 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2403500MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Bromfenac sodium hydrate (Bromfenac monosodium salt sesquihydrate) is the sodium salt form of bromfenac, a nonsteroidal anti-inflammatory drug (NSAID) with analgesic and anti-inflammatory properties. Upon ophthalmic administration, bromfenac binds to and inhibits cyclooxygenase II (COX II), an enzyme that converts arachidonic acid to cyclic endoperoxides, which are prostaglandin (PG) precursors. By inhibiting PG formation, bromfenac prevents PG-induced inflammation, vasodilation, leukocytosis, disruption of the blood-aqueous humor barrier, increased vascular permeability, and elevated intraocular pressure (IOP). Purity 99.65%
Catalog No. 50-287-6083