Learn More
Cayman Chemical BromfenacsodIum salt 10mg
SDP
Supplier:  Cayman Chemical 2376310 MG
Encompass_Preferred
A COX-2 inhibitor (IC50 = 6.6 nM); selective over COX-1 (IC50 = 210 nM); inhibits PGE2 production in a rabbit model of LPS-induced eye inflammation; decreases inflammation following cataract removal in dogs (0.9% topically on a tapering schedule for 24 weeks)
Catalog No. 50-296-4433