Learn More
Apexbio Technology LLC Bromhexine HCl 611-75-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18995.1
Encompass_Preferred
Bromhexine Hydrochloride is an expectorant compound utilized frequently in respiratory research to examine mucolytic mechanisms It functions primarily by disrupting acidic polysaccharide fibers within bronchial mucus reducing mucus viscosity and facilitating mucociliary clearance The compound is routinely employed in in vitro studies involving respiratory epithelial cells and ex vivo respiratory tract tissues to investigate its pharmacological influence on mucus production and secretion Reported IC50 values for Bromhexine Hydrochloride in mucolytic assays typically range between 30-50 M under standard experimental conditions Its applications extend to the study of respiratory disorders associated with abnormal mucus secretion enabling researchers to better characterize molecular pathways involved in mucus regulation and investigate potential therapeutic targets
Catalog No. 50-000-63763
 Restricted Item