Learn More
Medchemexpress LLC Bromhexine hydrochloride | 611-75-6 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0372A10MM/1ML
Encompass_Preferred
Bromhexine hydrochloride is identified as a potent and specific inhibitor of the TMPRSS2 protease, exhibiting an IC50 of 0.75 μM. It has shown potential in preventing and managing SARS-CoV-2 infection and is also recognized as an autophagy agonist. Additionally, it functions as a mucolytic cough suppressant, suggesting its utility in addressing a range of respiratory conditions.
  • Potent and specific TMPRSS2 protease inhibitor (IC50 of 0.75 μM).
  • Can prevent and manage SARS-CoV-2 infection.
  • Autophagy agonist.
  • Mucolytic cough suppressant with potential for various respiratory conditions.
Catalog No. 50-004-07228