Learn More
Medchemexpress LLC Bromhexine hydrochloride | 611-75-6 | 99.93% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0372A500MG
Encompass_Preferred
Bromhexine hydrochloride is a potent and specific TMPRSS2 protease inhibitor with an IC50 of 0.75 μM. It can prevent and manage SARS-CoV-2 infection. Bromhexine hydrochloride is also an autophagy agonist and a mucolytic cough suppressant, with potential for treating various respiratory conditions.
  • Potent and specific TMPRSS2 protease inhibitor
  • Can prevent and manage SARS-CoV-2 infection
  • Functions as an autophagy agonist
  • Serves as a mucolytic cough suppressant
  • Potential for treating various respiratory conditions
Catalog No. 50-004-06501