Learn More
ERA WATERS CORP Bromide - 1000mg/l, 12ml
SDP
Supplier:  ERA WATERS CORP 046
Encompass
One 125ml standard containing Bromide at 1000mg/l in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.
Catalog No. NC9167316