Learn More
Medchemexpress LLC Bromindione | 1146-98-1 | 98.11% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB091750MG
Encompass_Preferred
Bromindione is a potent and long-acting, Inandione-derived, oral anticoagulant compound. It is for research use only and not sold to patients.
  • Potent and long-acting anticoagulant
  • Inandione-derived compound
  • For research use only
Catalog No. 50-203-1142