Learn More
Sigma Aldrich Fine Chemicals Biosciences Bromine reagent grade100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2078884X100G
Encompass
Bromine is present in gaseous form in lower atmosphere generated by marine activity and as a by product produced from the leaded gasoline combustion. It is widely used as a brominating agent.
Catalog No. 50-289-7824