Learn More
Medchemexpress LLC Bromo-PEG1-C2-azide | 1144106-65-9 | 99.45% | C4H8BrN3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14081950MG
Encompass_Preferred
Bromo-PEG1-C2-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It also serves as a click chemistry reagent, featuring an azide group.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
Catalog No. 50-003-53799