Learn More
Medchemexpress LLC Bromo-PEG1-CH2COOH | 1135131-50-8 | ≥97.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1301635G
Encompass_Preferred
Bromo-PEG1-CH2COOH is a PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands joined by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs.
  • Composed of two distinct ligands joined by a linker.
  • One ligand targets an E3 ubiquitin ligase, the other targets the protein of interest.
  • Utilizes the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-003-38595