Learn More
Medchemexpress LLC Bromo-PEG2-alcohol | 57641-66-4 | 98.98% | C4H9BrO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1412135G
Encompass_Preferred
Bromo-PEG2-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53975