Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromo-PEG2-alcohol | 57641-66-4 | 99.0% | C4H9BrO2 | 10 G
SDP

Supplier:  Medchemexpress LLC HY14121310G

Encompass_Preferred

Bromo-PEG2-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed to exploit the intracellular ubiquitin-proteasome system, selectively degrading target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Molecular weight: 169.02
  • Chemical formula: C4H9BrO2
  • CAS number: 57641-66-4
  • Purity: 98.98%
  • Appearance: Liquid
  • Color: Colorless to light yellow

Catalog No. 50-003-53974


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.