Learn More
Medchemexpress LLC Bromo-PEG2-bromide | 31255-10-4 | 99.3% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1305895G
Encompass_Preferred
Bromo-PEG2-bromide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, and they facilitate the selective degradation of target proteins through the intracellular ubiquitin-proteasome system. It is for research use only.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Facilitates selective degradation of target proteins
  • Consists of two different ligands connected by a linker
  • For research use only
Catalog No. 50-003-40216