Learn More
Medchemexpress LLC Bromo-PEG2-phosphonic acid diethyl ester | 1226767-94-7 | 98% | C10H22BrO5P | 500 MG
Supplier: Medchemexpress LLC HY141299500MG
Bromo-PEG2-phosphonic acid diethyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This linker is essential for creating PROTACs, which degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein.
- Used in the synthesis of PROTACs
- PROTACs exploit the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins