Learn More
Medchemexpress LLC Bromo-PEG3-azide | 1446282-43-4 | 98.2% | C8H16BrN3O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140820100MG
Encompass_Preferred
Bromo-PEG4-azide is a PEG-based PROTAC linker. It is a click chemistry reagent that can be used in the synthesis of PROTACs. It contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-53362