Learn More
Medchemexpress LLC Bromo-PEG3-azide | 1446282-43-4 | 98.2% | C8H16BrN3O3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140820
Encompass_Preferred
Bromo-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also serves as a click chemistry reagent with an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • For research use only
Catalog No. 50-003-57260