Learn More
Medchemexpress LLC Bromo-peg3-azide | 1446282-43-4 | 98.2% | C8H16BrN3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14082050MG
Encompass_Preferred
Bromo-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent containing an azide group. PROTACs are utilized for the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-55501