Learn More
Medchemexpress LLC Bromo-PEG3-bromide | 31255-26-2 | 98.8% | C8H16Br2O3 | 10 G
SDP
Supplier:  Medchemexpress LLC HY14137310G
Encompass_Preferred
Bromo-PEG3-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Can be used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56307